MilliporeSigma
Potassium rhodizonate technical grade, 85%

Potassium rhodizonate

Potassium rhodizonate technical grade, 85%
Synonyms:
Rhodizonic acid potassium salt, 3,4,5,6-Tetraoxocyclohexene-1,2-diol dipotassium salt
Linear Formula:
C6O6K2
CAS Number:
Molecular Weight:
246.26
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Potassium rhodizonate, technical grade, 85%