MilliporeSigma
Probumin® Bovine Serum Albumin Universal Grade pkg of 10 g

Probumin® Bovine Serum Albumin Universal Grade

Probumin® Bovine Serum Albumin Universal Grade pkg of 10 g

Products

Product Category
Brand
Biological Source
Physical Form
Tissue
Quality Segment
Geographic Origin
Package Size
Technique

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-4 of 4