MilliporeSigma
Propyl isovalerate natural (US), ≥98%, FG

Propyl isovalerate

Propyl isovalerate natural (US), ≥98%, FG
Synonyms:
Propyl 3-methylbutanoate, Propyl 3-methylbutyrate
Linear Formula:
(CH3)2CHCH2COO(CH2)2CH3
CAS Number:
Molecular Weight:
144.21
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Propyl isovalerate, natural (US), ≥98%, FG