(S)-(−)-α-Ethylbenzylamine ≥95.0% (GC)

(S)-(−)-α-Ethylbenzylamine

(S)-(−)-α-Ethylbenzylamine ≥95.0% (GC)
Synonyms:
(S)-(−)-1-Phenylpropylamine
Empirical Formula (Hill Notation):
C9H13N
CAS Number:
Molecular Weight:
135.21

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
(S)-(−)-α-Ethylbenzylamine, ≥95.0% (GC)

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.