Skip to Content
MilliporeSigma
Sibiricose A5 ≥85% (LC/MS-ELSD)

Sibiricose A5

Sibiricose A5 ≥85% (LC/MS-ELSD)
Synonym(s):
3-O-Feruloylsucrose, Arillanin B
Empirical Formula (Hill Notation):
C22H30O14
CAS Number:
Molecular Weight:
518.47

Products

Shipping
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing