MilliporeSigma
SIGMAFAST™ p-Nitrophenyl phosphate Tablets tablet, To prepare 20 mL

SIGMAFAST p-Nitrophenyl phosphate Tablets

SIGMAFAST™ p-Nitrophenyl phosphate Tablets tablet, To prepare 20 mL
EC Number:

Products

Product Category
Brand
Physical Form
Quality Segment