Skip to Content
MilliporeSigma
Silica gel technical grade, pore size 60 Å, 230-400 mesh particle size, 40-63 μm particle size

Silica gel

Silica gel technical grade, pore size 60 Å, 230-400 mesh particle size, 40-63 μm particle size
CAS Number:
EC Number:

Products

Shipping
Product Category
Brand
Manufacturer
Physical Form
Purity
Quality Segment
Special Grade
Technique
Showing 1-20 of 38
Page 1 of 2
Page 1 of 2