Silver nanoplates resonant wavelength, 950 nm, 0.02 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized

Silver nanoplates

Silver nanoplates resonant wavelength, 950 nm, 0.02 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
Empirical Formula (Hill Notation):
Ag
Molecular Weight:
107.87

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
Silver nanoplates, resonant wavelength, 550 nm, 0.02 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
Silver nanoplates, resonant wavelength, 650 nm, 0.02 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
Silver nanoplates, resonant wavelength, 1050 nm, 0.02 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
Silver nanoplates, resonant wavelength, 750 nm, 0.02 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
Silver nanoplates, resonant wavelength, 850 nm, 0.02 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
Silver nanoplates, resonant wavelength, 950 nm, 0.02 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.