US
EN
Applications
Products
Services
Documents
Support
Home
Search Results
Sodium phosphate dibasic dihydrate
(5)
Sodium phosphate dibasic dihydrate
Synonyms:
sec
-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate
Linear Formula:
Na
2
HPO
4
· 2H
2
O
CAS Number:
10028-24-7
Molecular Weight:
177.99
EC Number:
231-448-7
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Showing 1-5 of 5
Page 1 of 1
Page 1 of 1
Compare
Product Number
Product Description
Pricing
71643
Sodium phosphate dibasic dihydrate,
BioUltra, for molecular biology, ≥99.0% (T)
Expand
View Pricing
30435
Sodium phosphate dibasic dihydrate,
puriss. p.a., reag. Ph. Eur., 98.5-101.0% (calc. to the dried substance)
Expand
View Pricing
04272
Sodium phosphate dibasic dihydrate,
meets analytical specification of Ph. Eur., BP, 98.5-101% (calc. to the dried substance)
Expand
View Pricing
71645
Sodium phosphate dibasic dihydrate,
BioXtra, ≥98.0% (T)
Expand
View Pricing
71662
Sodium phosphate dibasic dihydrate,
tested according to Ph. Eur.
Expand
View Pricing
Page 1 of 1
Page 1 of 1