MilliporeSigma
  • Home
  • Search Results
  • Sodium Phosphate, Dibasic, Molecular Biology Grade - CAS 7558-79-4 - Calbiochem (1)
Sodium Phosphate, Dibasic, Molecular Biology Grade - CAS 7558-79-4 - Calbiochem Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers.

Sodium Phosphate, Dibasic, Molecular Biology Grade - CAS 7558-79-4 - Calbiochem

Sodium Phosphate, Dibasic, Molecular Biology Grade - CAS 7558-79-4 - Calbiochem Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers.
Synonyms:
Sodium phosphate dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate
Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Sodium Phosphate, Dibasic, Molecular Biology Grade - CAS 7558-79-4 - Calbiochem, Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers.