US
EN
Applications
Products
Services
Documents
Support
Home
Search Results
Sodium rhodizonate dibasic
(1)
Sodium rhodizonate dibasic
Synonyms:
3,4,5,6-Tetraoxocyclohexene-1,2-diol disodium salt, Rhodizonic acid disodium salt
Empirical Formula (Hill Notation):
C
6
Na
2
O
6
CAS Number:
523-21-7
Molecular Weight:
214.04
EC Number:
208-340-3
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Showing 1-1 of 1
Page 1 of 1
Page 1 of 1
Compare
Product Number
Product Description
Pricing
R1609
Sodium rhodizonate dibasic,
97%
Expand
View Pricing
Page 1 of 1
Page 1 of 1