MilliporeSigma
Indium tin oxide ≥99.5% trace metals basis, 18 nm particle size (SEM), 20 wt. % in H2O

Indium tin oxide, dispersion

Indium tin oxide ≥99.5% trace metals basis, 18 nm particle size (SEM), 20 wt. % in H2O
Synonym(s):
ITO
CAS Number:

Products

Shipping
Brand
Boiling Point (°C)
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment