MilliporeSigma
Gentamicin sulfate British Pharmacopoeia (BP) Reference Standard

Gentamicin Sulfate

Gentamicin sulfate British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Gentamicin sulfate salt
CAS Number:

Products

Shipping
Brand
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique