MilliporeSigma
(+)-δ-3-Carene phyproof® Reference Substance

(+)-δ-3-Carene

(+)-δ-3-Carene phyproof® Reference Substance
Synonym(s):
(+)-3-Carene, (1S)-3,7,7-Trimethylbicyclo[4.1.0]hept-3-ene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23

Products

Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing