MilliporeSigma
Tamoxifen citrate British Pharmacopoeia (BP) Reference Standard

Tamoxifen Citrate

Tamoxifen citrate British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Tamoxifen citrate salt, (Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene
Empirical Formula (Hill Notation):
C26H29NO · C6H8O7
CAS Number:
Molecular Weight:
563.64

Products

Shipping
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique