MilliporeSigma
Titanium dioxide NIST® RM 8988, powder, particle size distribution

Titanium dioxide

Titanium dioxide NIST® RM 8988, powder, particle size distribution
Synonym(s):
Titanium(IV) oxide, Titania, Titanium dioxide
Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87

Products

Shipping
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique