MilliporeSigma
Phenol solution 100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Phenol

Phenol solution 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Synonym(s):
Hydroxybenzene
Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique