Skip to Content
MilliporeSigma
Stattic ≥98% (HPLC), powder

Stattic

Stattic ≥98% (HPLC), powder
Synonym(s):
6-Nitrobenzo[b]thiophene-1,1-dioxide, Stat three inhibitory compound, STAT3 Inhibitor V, Stattic
Empirical Formula (Hill Notation):
C8H5NO4S
CAS Number:
Molecular Weight:
211.19

Products

Shipping
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment