Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Sulindac sulfide ≥98% (HPLC), solid

Sulindac Sulfide

Sulindac sulfide ≥98% (HPLC), solid
Synonym(s):
Sulindac Sulfide, (Z)-5-Fluoro-2-methyl-1-[p-(methylthio)benzylidene]indene-3-acetic acid
Empirical Formula (Hill Notation):
C20H17FO2S
CAS Number:
Molecular Weight:
340.41
EC Number:

Products

Shipping
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing