MilliporeSigma
Thiamine for system suitability European Pharmacopoeia (EP) Reference Standard

Thiamine for system suitability

Thiamine for system suitability European Pharmacopoeia (EP) Reference Standard
Synonyms:
Thiamine hydrochloride, Aneurine hydrochloride, Vitamin B1 hydrochloride
Empirical Formula (Hill Notation):
C12H17ClN4OS · HCl
CAS Number:
Molecular Weight:
337.27

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Thiamine for system suitability, European Pharmacopoeia (EP) Reference Standard