Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now
Thiostrepton United States Pharmacopeia (USP) Reference Standard

Thiostrepton

Thiostrepton United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Thiostrepton
Empirical Formula (Hill Notation):
C72H85N19O18S5
CAS Number:
Molecular Weight:
1664.89

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Research Area
Quality Segment
Application
API Family
Reference Material Type