Skip to Content
MilliporeSigma
Titanium(IV) oxide, rutile <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm

Titanium dioxide, rutile

Titanium(IV) oxide, rutile <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Synonym(s):
Titanium dioxide
Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:

Products

Shipping
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Length (mm)
Quality Segment
Application
Particle Size (µm)
Reagent Type
Core