MilliporeSigma
trans-1,3-Diphenyl-2,3-epoxypropan-1-one ≥98.0%

trans-1,3-Diphenyl-2,3-epoxypropan-1-one

trans-1,3-Diphenyl-2,3-epoxypropan-1-one ≥98.0%
Synonyms:
trans-2-Benzoyl-3-phenyloxirane, trans-Benzalacetophenone oxide
Empirical Formula (Hill Notation):
C15H12O2
CAS Number:
Molecular Weight:
224.25

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing