MilliporeSigma
Trizma® hemisulfate

Trizma® hemisulfate

Trizma® hemisulfate
Synonyms:
Bis(tris[hydroxymethyl]aminomethane) sulfate, TRIS hemisulfate salt, Tris(hydroxymethyl)aminomethane hemisulfate salt
Linear Formula:
NH2C(CH2OH)3 · 0.5 H2SO4
CAS Number:
Molecular Weight:
170.17
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Trizma® hemisulfate