US
EN
Applications
Products
Services
Support
Home
Search Results
Trypsin-EDTA Solution 10X
(1)
Trypsin-EDTA Solution 10X
Synonyms:
Trypsin solution from porcine pancreas
CAS Number:
9002-07-7
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
59418C
Trypsin-EDTA Solution 10X
Expand
View Pricing