US
EN
Applications
Products
Services
Documents
Support
Home
Search Results
Tryptic Soy Agar
(20)
Tryptic Soy Agar
Synonyms:
Tryptic Soy Agar, CASO Agar, Soybean Casein digest Agar, TSA, Tryptone Soya Agar
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Showing 1-20 of 20
Page 1 of 1
Page 1 of 1
Compare
Product Number
Product Description
Pricing
1.46231
Tryptic Soy Agar,
TSA with Lecithin, Tween
®
, Histidine and Sodium thiosulfate - ICR Contact plate, irradiated, triple packed for environmental monitoring (Isolator and Clean room)
Expand
View Pricing
1.46552
Tryptic Soy Agar,
TSA with Lecithin and Tween
®
- ICR plus Contact plate, triple bagged, irradiated for environmental monitoring (Isolator and Clean room)
Expand
View Pricing
1.46001
Tryptic Soy Agar,
ICR 30ml
Expand
View Pricing
1.46069
Tryptic Soy Agar,
with Lecithin, Tween
®
, Histidine and Sodium thiosulfate- ICR 30ml
Expand
View Pricing
1.46685
Tryptic Soy Agar,
ICR+ 90 mm diameter,triple-bagged, irradiated with a locking lid for use in cleanroom environment.
Expand
View Pricing
1.46783
Tryptic Soy Agar,
TSA with Lecithin, Tween
®
, histidine and sodium thiosulfate- ICR plus Contact plate, irradiated, triple packed, for environmental monitoring (Isolator and Clean room)
Expand
View Pricing
22091
Tryptic Soy Agar,
NutriSelect
®
Plus, suitable for microbiology
Expand
View Pricing
1.46683
Tryptic Soy Agar,
with Lecithin, Tween
®
, Histidine and Sodium thiosulfate - ICR+ 90 mm diameter
Expand
View Pricing
1.46195
Tryptic Soy Agar,
TSA with Lecithin and Tween
®
- ICR Contact plate, irradiated, triple packed, for environmental monitoring (Isolator and Clean room)
Expand
View Pricing
1.46050
Tryptic Soy Agar,
with Lecithin and Tween
®
- ICR 30ml
Expand
View Pricing
1.46684
Tryptic Soy Agar,
with Lecithin and Tween
®
- ICR+ 90 mm diameter with a locking lid, for use with cleanroom environments
Expand
View Pricing
1.46431
Tryptic Soy Agar,
ReadyPlate
™
90mm plate in accordance with ISO, FDA-BAM and EP + USP for detection of wide range of bacteria by Membrane filtration method.
Expand
View Pricing
1.46429
Tryptic Soy Agar,
ready-to-use, tube volume 18 mL , Filling volume
Expand
View Pricing
146800
Tryptic Soy Agar,
with Penase, Lecithin, Tween
®
, Histidine and Sodium thiosulfate - ICR+ 30ml, diam. 90 mm
Expand
View Pricing
146798
Tryptic Soy Agar,
TSA with Penase, Lecithin, Tween®, Histidine and Sodium thiosulfate - ICR Contact plate, irradiated, triple packed for environmental monitoring (Isolator and Clean room), diam. 55 mm
Expand
View Pricing
1.46076
Tryptic Soy Agar,
with Lecithin, Tween
®
and Cephase- ICR 30ml
Expand
View Pricing
1.46778
Tryptic Soy Agar,
with Lecithin, Tween
®
, Histidine and Sodium thiosulfate - ICR 15 cm diameter
Expand
View Pricing
1.46700
Tryptic Soy Agar,
with Cephase, Lecithin and Tween
®
- ICR+ 90 mm diameter
Expand
View Pricing
1.46820
Tryptic Soy Agar,
TSA with Lecithin, Tween
®
, Histidine and Sodium thiosulfate – 15cm ICR Settle plate, irradiated, triple packed, for environmental monitoring (Isolator and Clean room)
Expand
View Pricing
146799
Tryptic Soy Agar,
with Penase, Lecithin, Tween
®
, Histidine and Sodium thiosulfate - ICR 30ml, diam. 90 mm
Expand
View Pricing
Page 1 of 1
Page 1 of 1