MilliporeSigma
Tryptic Soy Broth (TSB) bottle volume 250 mL , filling volume

Tryptic Soy Broth(TSB)

Tryptic Soy Broth (TSB) bottle volume 250 mL , filling volume

Products

Product Category
Brand
Physical Form
Sterilization Compatibility
Agency
Capacity (mL)
Volume (ml)
Quality Segment
Closure Type
Application
Bottle material
Vial Bottle Material
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique