
Tryptic Soy Broth with Neutralizers

Synonym(s):
CASO, Casein Soybean Digest Broth with 3% Tween, 0.3% Lecithin, 0.1% Histidine and 0.1% Cysteine, SCDB, Tryptic Soy Broth with TLHC (Inactivator B.), Tryptic Soy Broth with neutralizer
Products
Product Category
Brand
Color
Physical Form
Capacity (mL)
Volume (ml)
Quality Segment
Application
Sterility
Technique
Showing 1-13 of 13
Compare
Product Number
Description
Pricing