US
EN
Applications
Products
Services
Documents
Support
Home
Search Results
TSKgel
®
SP-STAT HPLC Column
(2)
TSKgel
®
SP-STAT HPLC Column
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Showing 1-2 of 2
Page 1 of 1
Page 1 of 1
Compare
Product Number
Product Description
Pricing
821964
TSKgel
®
SP-STAT HPLC Column,
phase sulfonic acid, L × I.D. 10 cm × 4.6 mm, 7 μm particle size
Expand
View Pricing
821963
TSKgel
®
SP-STAT HPLC Column,
phase sulfonic acid, L × I.D. 3.5 cm × 3 mm, 10 μm particle size
Expand
View Pricing
Page 1 of 1
Page 1 of 1