Skip to Content
MilliporeSigma
ZM 241385 ≥98% (HPLC)

ZM 241385

ZM 241385 ≥98% (HPLC)
Synonym(s):
4-(-2-[7-amino-2-{2-furyl}{1,2,4}triazolo{2,3-a} {1,3,5}triazin-5-yl-amino]ethyl)phenol, Adenosine A2A Receptor Antagonist I, ZM 241385
Empirical Formula (Hill Notation):
C16H15N7O2
CAS Number:
Molecular Weight:
337.34

Products

Shipping
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Showing 1-2 of 2