US
EN
Applications
Products
Services
Support
Home
Search Results
Z-Tyr-OH
(1)
Z-Tyr-OH
Synonyms:
N
-Carbobenzyloxy-L-tyrosine, Z-L-Tyrosine
Linear Formula:
HOC
6
H
4
CH
2
CH(NHCOOCH
2
C
6
H
5
)COOH
CAS Number:
1164-16-5
Molecular Weight:
315.32
EC Number:
214-609-6
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
470597
Z-Tyr-OH, 97%
Expand
View Pricing