  • Search Results
  • Rapid analysis of parathion in biological samples using headspace solid-phase micro-extraction (HS-SPME) and gas chromatography/mass spectrometry (GC/MS).
Rapid analysis of parathion in biological samples using headspace solid-phase micro-extraction (HS-SPME) and gas chromatography/mass spectrometry (GC/MS).
Clinical chemistry and laboratory medicine (1999-09-04)
F Musshoff, H Junker, B Madea
ABSTRACT
A simple and rapid method for the analysis of parathion in biological samples is presented. The method consists of the extraction of parathion from blood samples by headspace solid-phase micro-extraction (SPME), followed by capillary gas chromatography and mass spectrometry detection. The recoveries in the blood samples after addition of ammonium sulphate and sulphuric acid were between 85% and 89% compared to samples prepared in water. Linearity was established over a concentration range of 0.1-5 microg/g blood with acceptable coefficients of correlation and limits of detection reached 0.02-0.05 microg/g. The time for an analysis is 57 minutes for one sample, including the extraction step. In conclusion, HS-SPME in combination with GC/MS is an effective method for the determination and quantification of parathion-ethyl and parathion-methyl in biological material.
Product Description
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 1 cm
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm (CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with autosampler, needle size 24 ga
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm (CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm (CAR/PDMS), needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm (CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder/autosampler, fiber L 2 cm
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), for use with manual holder, needle size 24 ga
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with manual holder/autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm (PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm (PDMS), needle size 24 ga, for use with manual holder