  • Search Results
  • Analysis of the volatile compounds of flowers and essential oils from Lavandula angustifolia cultivated in Northeastern Italy by headspace solid-phase microextraction coupled to gas chromatography-mass spectrometry.
Analysis of the volatile compounds of flowers and essential oils from Lavandula angustifolia cultivated in Northeastern Italy by headspace solid-phase microextraction coupled to gas chromatography-mass spectrometry.
Planta medica (2008-02-06)
Carla Da Porto, Deborha Decorti
ABSTRACT
Headspace solid-phase microextraction (HS-SPME) coupled to gas chromatography and mass spectrometry using the divinyl/carboxen/polydimethylsiloxane (DVB-CAR-PDMS) fibre was applied for the analysis of aroma profiles of Lavandula angustifolia L. flowers and the corresponding essential oils. The optimal sampling time was determined by studying the equilibrium time profile of the major volatile compounds for the lavender flowers (50 min) and the essential oil (20 min). Comparative analysis of L. Angustifolia L. cultivated in Friuli Venezia Giulia (northeastern Italy) highlighted that the contents of linalool and linalyl acetate were the major differences between the composition of flowers and the hydro-distilled products. Lavender essential oil from Middle-Friuli Venezia Giulia was evaluated as the highest quality for its high level of linalyl acetate (31.7 %) and linalool (45.0 %) and low percentage of camphor (0.5 %). The use of headspace SPME was shown to be a convenient and effective analytical tool for the sampling of volatile compounds and it could be used to test the quality of flowers and essential oils from Lavandula species.
Product Description
Supelco
Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4  in. × 3 1/2  in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Supelco
ORBO 78 HBr on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
Supelco
Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: (300 mg), cartridge volume 3 mL, pkg of 10 ea
Supelco
ORBO-93 Sorbent Tube, matrix Carboxen® 1000, ORBO tube I.D. × L 6 mm × 95 mm, Bed B 90 mg, Bed A 180 mg, 60/80 mesh, pkg of 25 ea
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
Carboxen® 569, O.D. × L 1/4  in. × 3 1/2 , glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: 300 mg, cartridge size 3 mL, pkg of 50 ea
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 23 ga, for use with manual holder
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with manual holder/autosampler
Supelco
Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder/autosampler, fiber L 2 cm
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm (CAR/PDMS), needle size 24 ga, for use with manual holder, StableFlex fiber