  • Search Results
  • Needle-trap device for the sampling and determination of chlorinated volatile compounds.
Needle-trap device for the sampling and determination of chlorinated volatile compounds.
Journal of separation science (2013-08-21)
Elżbieta Dobrzyńska, Bogusław Buszewski
ABSTRACT
A needle-trap device, with immobilized sorbent inside the syringe, coupled with GC-MS was applied for air sampling and determination of chlorinated volatile organic compounds such as dichloromethane, trichloromethane, and tetrachloromethane. The application of a needle trap packed with combination of three sorbents including Tenax TA, Carbopack X, and Carboxen 1000 resulted in detection limits of few pg for chlorinated volatile compounds and recoveries of 99.2-102.8%. The extraction and desorption parameters were optimized within the study. As a result, the precision determined as RSD was equal to 5.05 and 3.03 and 6.52% for dichloromethane, trichloromethane, and tetrachloromethane, respectively. The storage time for chlorinated compounds up to 48 h and reusability of the needle-trap device were verified. The obtained results have proved the ability of needle traps to compete with other solventless sampling and sample preparation extraction techniques.
Product Description
Supelco
Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4  in. × 3 1/2  in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Supelco
ORBO 78 HBr on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
Carbopack Adsorbent, matrix Carbopack B, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carbopack Adsorbent, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
Carbopack Adsorbent, matrix Carbopack Z, 60-80 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
Supelco
Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: (300 mg), cartridge volume 3 mL, pkg of 10 ea
Supelco
ORBO-93 Sorbent Tube, matrix Carboxen® 1000, ORBO tube I.D. × L 6 mm × 95 mm, Bed B 90 mg, Bed A 180 mg, 60/80 mesh, pkg of 25 ea
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
Carboxen® 569, O.D. × L 1/4  in. × 3 1/2 , glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: 300 mg, cartridge size 3 mL, pkg of 50 ea
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 23 ga, for use with manual holder
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with manual holder/autosampler
Supelco
Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (CAR/PDMS), needle size 24 ga, for use with autosampler