  • Search Results
  • Paired box 2 upregulates androgen receptor gene expression in androgen-independent prostate cancer.
Paired box 2 upregulates androgen receptor gene expression in androgen-independent prostate cancer.
The FEBS journal (2014-08-19)
Saya Ito, Takashi Ueda, Akihisa Ueno, Hideo Nakagawa, Hidefumi Taniguchi, Fumiya Hongo, Kazumi Kamoi, Koji Okihara, Akihiro Kawauchi, Tsuneharu Miki
ABSTRACT
Androgen-independent prostate cancer is known as a hormone-refractory disease. Although the androgen receptor (AR) is considered to be a key regulator of androgen-independent prostate cancer progression, the mechanism through which AR gene expression is regulated is not well understood. In the present study, we showed that the AR gene was upregulated by paired box 2 (PAX2) in androgen-independent prostate cancer. When PAX2 upregulated AR gene expression, a decrease in DNA methylation of the AR gene locus was also observed. PAX2 was highly expressed and promoted cell growth in an androgen-independent prostate cancer cell line (22Rv1). The cell growth inhibition by PAX2 knockdown was rescued by AR overexpression in 22Rv1 cells. In a mouse xenograft model of androgen-independent prostate cancer, PAX2 knockdown inhibited tumor growth and AR gene expression and also increased DNA methylation of the AR gene. Consistent with this, AR and PAX2 expression levels were positively correlated in prostate cancer patients. These findings suggested that PAX2 promoted cancer cell growth in androgen-independent prostate cancer by regulating AR gene expression through an epigenetic mechanism.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder