  • Search Results
  • Neutral oligosaccharides in the urine of a patient with glycogen storage disease type II.
Neutral oligosaccharides in the urine of a patient with glycogen storage disease type II.
Journal of biochemistry (1985-10-01)
M Kuriyama, R Hiwatari, T Ariga, Y Sakano, J Abe, M Osame, A Igata
ABSTRACT
Neutral oligosaccharides were isolated from urine of an adult patient with glycogen storage disease type II, a deficiency of lysosomal acid alpha-glucosidase, by chromatography on columns of activated charcoal, Dowex 50 X 2 and Dowex 1 X 2. Total neutral oligosaccharides in the urine of the patient were increased about 5-fold as compared with those in normal controls. The most accumulated oligosaccharide was separated by Bio-Gel P-2 column chromatography, and finally purified by paper chromatography. Based on various studies, including carbohydrate analysis, chemical ionization mass spectrometry, fast atom bombardment mass spectrometry, degradation by glucoamylase and isopullulanase, and methylation analysis, the structure of this oligosaccharide was deduced to be Glc alpha 1----6Glc alpha 1----4Glc alpha 1----4Glc. This oligosaccharide appears to be accumulated in urine of the patient with acid alpha-glucosidase deficiency as an end product of the hydrolysis of glycogen.
Sigma-Aldrich
Octadecylthioethyl 4-O-(4-O-[6-O-α-D-glucopyranosyl-α-D-glucopyranosyl]-α-D-glucopyranosyl)-β-D-glucopyranoside, ≥95%