US
EN
Applications
Products
Services
Support
Home
Search Results
Trityl tetrakis [3, 5-bis (trifluoromethyl) phenyl] borate: a new hydride abstraction reagent.
Trityl tetrakis [3, 5-bis (trifluoromethyl) phenyl] borate: a new hydride abstraction reagent.
The Journal of Organic Chemistry (1992)
Bahr S R and Boudjouk P