[(3, 5-(CF3) 2C6H3) 4B]-[H (OEt2) 2]+: a convenient reagent for generation and stabilization of cationic, highly electrophilic organometallic complexes.
[(3, 5-(CF3) 2C6H3) 4B]-[H (OEt2) 2]+: a convenient reagent for generation and stabilization of cationic, highly electrophilic organometallic complexes.