  • Search Results
  • PRMT1 negatively regulates activation-induced cell death in macrophages by arginine methylation of GAPDH.
PRMT1 negatively regulates activation-induced cell death in macrophages by arginine methylation of GAPDH.
Experimental cell research (2018-04-18)
Jun-Ho Cho, Rana Lee, Eunju Kim, Yea Eun Choi, Eui-Ju Choi
ABSTRACT
Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is implicated in cell death in addition to a role as a glycolytic enzyme. In particular, when cells are exposed to cellular stressors involving nitric oxide (NO) production, GAPDH can undergo NO-induced S-nitrosylation and S-nitrosylated GAPDH has been shown to elicit apoptosis. However, the mechanism underlying the regulation of the pro-apoptotic function of GAPDH remains unclear. Here, we found that protein arginine methyltransferase 1 (PRMT1) mediated arginine methylation of GAPDH in primary bone marrow-derived macrophages in a NO-dependent manner. Moreover, PRMT1 inhibited S-nitrosylation of GAPDH as well as its binding to SIAH1, thereby reducing the nuclear translocation of GAPDH in lipopolysaccharide (LPS)/interferon (IFN)-γ-activated macrophages. Furthermore, depletion of PRMT1 expression by RNA interference potentiated LPS/IFN-γ-induced apoptosis in macrophages. Taken together, our results suggest that PRMT1 has a previously unrecognized function to inhibit activation-induced cell death of macrophages through arginine methylation of GAPDH.
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
S-Nitrosoglutathione, ≥97%
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat