  • Search Results
  • 3D Microcontact Printing for Combined Chemical and Topographical Patterning on Porous Cell Culture Membrane.
3D Microcontact Printing for Combined Chemical and Topographical Patterning on Porous Cell Culture Membrane.
ACS applied materials & interfaces (2018-06-09)
Justyna Borowiec, Joerg Hampl, Sukhdeep Singh, Sebastian Haefner, Karin Friedel, Patrick Mai, Dana Brauer, Florian Ruther, Liliana Liverani, Aldo R Boccaccini, Andreas Schober
ABSTRACT
Micrometer-scale biochemical or topographical patterning is commonly used to guide the cell attachment and growth, but the ability to combine these patterns into an integrated surface with defined chemical and geometrical characteristics still remains a technical challenge. Here, we present a technical solution for simultaneous construction of 3D morphologies, in the form of channels, on porous membranes along with precise transfer of extracellular matrix proteins into the channels to create patterns with geometrically restricting features. By combining the advantages of microthermoforming and microcontact printing, this technique offers a unique patterning process that provides spatiotemporal control over morphological and chemical feature in a single step. By use of our 3D-microcontact printing (3DμCP), determined microstructures like channels with different depths and widths even with more complex patterns can be fabricated. Collagen, fibronectin, and laminin were successfully transferred inside the predesigned geometries, and the validity of the process was confirmed by antibody staining. Cells cultivated on 3DμCP patterned polycarbonate membrane have shown selective adhesion and growth. This technique offers a novel tool for creating freeform combinatorial patterning on the thermoformable surface.
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution