  • Search Results
  • Immune response induced by major environmental pollutants through altering neutrophils in zebrafish larvae.
Immune response induced by major environmental pollutants through altering neutrophils in zebrafish larvae.
Aquatic toxicology (Amsterdam, Netherlands) (2018-06-15)
Hongyan Xu, Xiaoyan Zhang, Hankun Li, Caixia Li, Xiao-Jing Huo, Li-Ping Hou, Zhiyuan Gong
ABSTRACT
Environmental pollutants may cause adverse effects on the immune system of aquatic organisms. However, the cellular effects of pollutants on fish immune system are largely unknown. Here, we exploited the transgenic zebrafish Tg(lysC:DsRed2) larva as a preliminary screening system to evaluate the potential inflammatory effects of environmental pollutants. Tg(lysC:DsRED2) larvae aged 7-day-postfertilization (7 dpf) were treated with selected environmental chemicals for 24 h (24 h) and the number of neutrophils were quantified using both image analysis and fluorescence activated cell sorting (FACS). We found that the numbers of neutrophils in the Tg(lysC:DsRED2) larvae were significantly increased by most of the organic chemicals tested, including E2 (17β-estradiol), BPA (Bisphenol-A), NDEA (N-nitrosodiethylamine), 4-NP (4-Nitrophenol) and Lindane (γ-hexachlorocyclohexane). Neutrophil numbers were also increased by all the metals tested (Na2HAsO4· 7H2O, Pb(NO3)2, HgCl2, CdCl2, CuSO4·5H2O, ZnSO4, and K2Cr2O7). The only exception was TCDD (2,3,7,8-tetrachlorodibenzo-p-dioxin), which significantly reduced the number of neutrophils after exposure. Additionally, the transcription of genes (lyz, mpo, tnfα and il8) related to fish immune system were significantly modulated upon exposure to some of the selected chemicals such as E2, TCDD, Cu and Cd. This study revealed that representatives of major categories of environmental pollutants could cause an acute inflammatory response in zebrafish larvae as shown by alterations in the neutrophils, which may imply a common immunotoxicity mechanism for most environmental pollutants. This study has also demonstrated that Tg(lyz:DsRed2) transgenic zebrafish is an excellent tool for screening environmental chemicals with potential inflammatory effects through FACS-facilitated neutrophil counting.
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein