  • Search Results
  • The ubiquitin ligase TRIM56 inhibits ovarian cancer progression by targeting vimentin.
The ubiquitin ligase TRIM56 inhibits ovarian cancer progression by targeting vimentin.
Journal of Cellular Physiology (2018)
Zhao L, et al.