  • Search Results
  • Influence of curing protocol and ceramic composition on the degree of conversion of resin cement.
Influence of curing protocol and ceramic composition on the degree of conversion of resin cement.
Journal of applied oral science : revista FOB (2017-12-07)
Marcos Daniel Septimio Lanza, Marcello Rubens Barsi Andreeta, Thiago Amadei Pegoraro, Luiz Fernando Pegoraro, Ricardo Marins De Carvalho
ABSTRACT
Due to increasing of aesthetic demand, ceramic crowns are widely used in different situations. However, to obtain long-term prognosis of restorations, a good conversion of resin cement is necessary. To evaluate the degree of conversion (DC) of one light-cure and two dual-cure resin cements under a simulated clinical cementation of ceramic crowns. Prepared teeth were randomly split according to the ceramic's material, resin cement and curing protocol. The crowns were cemented as per manufacturer's directions and photoactivated either from occlusal suface only for 60 s; or from the buccal, occlusal and lingual surfaces, with an exposure time of 20 s on each aspect. After cementation, the specimens were stored in deionized water at 37°C for 7 days. Specimens were transversally sectioned from occlusal to cervical surfaces and the DC was determined along the cement line with three measurements taken and averaged from the buccal, lingual and approximal aspects using micro-Raman spectroscopy (Alpha 300R/WITec®). Data were analyzed by 3-way ANOVA and Tukey test at =5%. Statistical analysis showed significant differences among cements, curing protocols and ceramic type (p<0.001). The curing protocol 3x20 resulted in higher DC for all tested conditions; lower DC was observed for Zr ceramic crowns; Duolink resin cement culminated in higher DC regardless ceramic composition and curing protocol. The DC of resin cement layers was dependent on the curing protocol and type of ceramic.
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block