  • Search Results
  • Effect of Insufficient Light Exposure on Polymerization Kinetics of Conventional and Self-adhesive Dual-cure Resin Cements.
Effect of Insufficient Light Exposure on Polymerization Kinetics of Conventional and Self-adhesive Dual-cure Resin Cements.
Operative dentistry (2016-10-01)
Y Jang, J L Ferracane, C S Pfeifer, J W Park, Y Shin, B D Roh
ABSTRACT
The purpose of this study was to investigate the influence of insufficient light exposure on the polymerization of conventional and self-adhesive dual-cure resin cements under ceramic restorations. Two conventional dual-cure resin cements (Rely-X ARC, Duolink) and two self-adhesive resin cements (Rely-X U200, Maxcem Elite) were polymerized under different curing modes (dual-cure or self-cure), curing times (20 and 120 seconds), and thickness of a ceramic overlay (2 and 4 mm). Polymerization kinetics was measured by Fourier transform infrared spectroscopy for the initial 10 minutes and after 24 hours. Data were analyzed using mixed model analysis of variance (ANOVA), one-way ANOVA/Student-Newman-Keuls post hoc test, and paired t-test (α=0.05). When light-curing time was set to 20 seconds, the presence of the ceramic block significantly affected the degree of conversion (DC) of all resin cements. Especially, the DC of the groups with 20 seconds of light-curing time under 4 mm of ceramic thickness was even lower than that of the self-cured groups at 24 hours after polymerization (p<0.05). However, when light-curing time was set to 120 seconds, a similar DC compared with the group with direct light exposure (p>0.05) was achieved in all dual-cure groups except Maxcem Elite, at 24 hours after polymerization. For both conventional and self-adhesive dual-cure resin cements, insufficient light exposure (20 seconds of light-curing time) through thick ceramic restoration (4 mm thick) resulted in a DC even lower than that of self-curing alone.
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block