  • Search Results
  • Maturation and processing of the amyloid precursor protein is regulated by the potassium/sodium hyperpolarization-activated cyclic nucleotide-gated ion channel 2 (HCN2).
Maturation and processing of the amyloid precursor protein is regulated by the potassium/sodium hyperpolarization-activated cyclic nucleotide-gated ion channel 2 (HCN2).
Biochemical and biophysical research communications (2016-12-27)
Susanne Frykman, Mitsuhiro Inoue, Atsushi Ikeda, Yasuhiro Teranishi, Takahiro Kihara, Jolanta L Lundgren, Natsuko G Yamamoto, Nenad Bogdanovic, Bengt Winblad, Sophia Schedin-Weiss, Lars O Tjernberg
ABSTRACT
The toxic amyloid β-peptide (Aβ) is a key player in Alzheimer Disease (AD) pathogenesis and selective inhibition of the production of this peptide is sought for. Aβ is produced by the sequential cleavage of the Aβ precursor protein (APP) by β-secretase (to yield APP-C-terminal fragment β (APP-CTFβ) and soluble APPβ (sAPPβ)) and γ-secretase (to yield Aβ). We reasoned that proteins that associate with γ-secretase are likely to regulate Aβ production and to be targets of pharmaceutical interventions and therefore performed a pull-down assay to screen for such proteins in rat brain. Interestingly, one of the purified proteins was potassium/sodium hyperpolarization-activated cyclic nucleotide-gated ion channel 2 (HCN2), which has been shown to be involved in epilepsy. We found that silencing of HCN2 resulted in decreased secreted Aβ levels. To further investigate the mechanism behind this reduction, we also determined the levels of full-length APP, sAPP and APP-CTF species after silencing of HCN2. A marked reduction in sAPP and APP-CTF, as well as glycosylated APP levels was detected. Decreased Aβ, sAPP and APP-CTF levels were also detected after treatment with the HCN2 inhibitor ZD7288. These results indicate that the effect on Aβ levels after HCN2 silencing or inhibition is due to altered APP maturation or processing by β-secretase rather than a direct effect on γ-secretase. However, HCN2 and γ-secretase were found to be in close proximity, as evident by proximity ligation assay and immunoprecipitation. In summary, our results indicate that silencing or inhibition of HCN2 affects APP processing and thereby could serve as a potential treatment strategy.
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat