  • Search Results
  • Evaluation of Radiopacity of Luting Cements Submitted to Different Aging Procedures.
Evaluation of Radiopacity of Luting Cements Submitted to Different Aging Procedures.
Journal of prosthodontics : official journal of the American College of Prosthodontists (2018-10-16)
Rafael B Junqueira, Rodrigo F Carvalho, Felipe A G F Yamamoto, Solange M Almeida, Francielle S Verner
ABSTRACT
To evaluate the radiopacity of luting cements submitted to different aging procedures. Twenty discs (1 × 4 mm) of each of the following cements were prepared: zinc phosphate, RelyX Luting 2, Variolink 2, AllCem, RelyX U200, Multilink, Panavia F2.0, and RelyX ARC. Then, they were randomly divided into two groups (n = 10/group), according to the aging procedure: thermal cycling group and water storage group. Before and after aging procedures, specimens were x-rayed with an aluminum step-wedge (11 steps, 1 mm thick each) and three dental slices (1 mm thick). The radiopacity was evaluated by means of optical density, which was measured using ImageJ software. The values were converted into millimeters of aluminum with a logistic regression calibration curve. The data were analyzed by one-way ANOVA and Tukey post-hoc test (α = 0.05). In the thermal cycling group, RelyX Luting 2, RelyX U200, and Panavia F2.0 showed a statistically significant reduction in radiopacity (p < 0.05). In the water storage group, all cements showed a significant reduction in radiopacity (p < 0.05), and RelyX Luting 2 and Panavia F2.0 performed below the desired ISO 4049 standard. The radiopacity of luting cements can decrease after aging, especially after 1-year water storage.
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block