  • Search Results
  • MicroRNA-10a is crucial for endothelial response to different flow patterns via interaction of retinoid acid receptors and histone deacetylases.
MicroRNA-10a is crucial for endothelial response to different flow patterns via interaction of retinoid acid receptors and histone deacetylases.
Proceedings of the National Academy of Sciences of the United States of America (2017-02-09)
Ding-Yu Lee, Ting-Er Lin, Chih-I Lee, Jing Zhou, Yi-Hsuan Huang, Pei-Ling Lee, Yu-Tsung Shih, Shu Chien, Jeng-Jiann Chiu
ABSTRACT
Histone deacetylases (HDACs) and microRNAs (miRs) have emerged as two important epigenetic factors in the regulation of vascular physiology. This study aimed to elucidate the relationship between HDACs and miRs in the hemodynamic modulation of endothelial cell (EC) dysfunction. We found that miR-10a has the lowest expression among all examined shear-responsive miRs in ECs under oscillatory shear stress (OS), and a relatively high expression under pulsatile shear stress (PS). PS and OS alter EC miR-10a expression to regulate the expression of its direct target GATA6 and downstream vascular cell adhesion molecule (VCAM)-1. PS induces the expression, nuclear accumulation, and association of retinoid acid receptor-α (RARα) and retinoid X receptor-α (RXRα). RARα and RXRα serve as a "director" and an "enhancer," respectively, to enhance RARα binding to RA-responsive element (RARE) and hence miR-10a expression, thus down-regulating GATA6/VCAM-1 signaling in ECs. In contrast, OS induces associations of "repressors" HDAC-3/5/7 with RARα to inhibit the RARα-directed miR-10a signaling. The flow-mediated miR-10a expression is regulated by Krüppel-like factor 2 through modulation in RARα-RARE binding, with the consequent regulation in GATA6/VCAM-1 in ECs. These results are confirmed in vivo by en face staining on the aortic arch vs. the straight thoracic aorta of rats. Our findings identify a mechanism by which HDACs and RXRα modulate the hormone receptor RARα to switch miR-10a expression and hence the proinflammatory vs. anti-inflammatory responses of vascular endothelium under different hemodynamic forces.
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat