  • Search Results
  • Bicaudal D2 facilitates the cytoplasmic trafficking and nuclear import of HIV-1 genomes during infection.
Bicaudal D2 facilitates the cytoplasmic trafficking and nuclear import of HIV-1 genomes during infection.
Proceedings of the National Academy of Sciences of the United States of America (2017-11-29)
Adarsh Dharan, Silvana Opp, Omar Abdel-Rahim, Sevnur Komurlu Keceli, Sabrina Imam, Felipe Diaz-Griffero, Edward M Campbell
ABSTRACT
Numerous viruses, including HIV-1, exploit the microtubule network to traffic toward the nucleus during infection. Although numerous studies have observed a role for the minus-end microtubule motor dynein in HIV-1 infection, the mechanism by which the viral core containing the viral genome associates with dynein and induces its perinuclear trafficking has remained unclear. Here, we report that the dynein adapter protein bicaudal D2 (BICD2) is able to interact with HIV-1 viral cores in target cells. We also observe that BICD2 can bind in vitro-assembled capsid tubes through its CC3 domain. We observe that BICD2 facilitates infection by promoting the trafficking of viral cores to the nucleus, thereby promoting nuclear entry of the viral genome and infection. Finally, we observe that depletion of BICD2 in the monocytic cell line THP-1 results in an induction of IFN-stimulated genes in these cells. Collectively, these results identify a microtubule adapter protein critical for trafficking of HIV-1 in the cytoplasm of target cells and evasion of innate sensing mechanisms in macrophages.
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat