  • Search Results
  • Conclusion regarding the peer review of the pesticide risk assessment of the active substance Dimoxystrobin.
Conclusion regarding the peer review of the pesticide risk assessment of the active substance Dimoxystrobin.
EFSA Journal (2005)