  • Search Results
  • Identification of GPC2 as an Oncoprotein and Candidate Immunotherapeutic Target in High-Risk Neuroblastoma.
Identification of GPC2 as an Oncoprotein and Candidate Immunotherapeutic Target in High-Risk Neuroblastoma.
Cancer cell (2017-09-13)
Kristopher R Bosse, Pichai Raman, Zhongyu Zhu, Maria Lane, Daniel Martinez, Sabine Heitzeneder, Komal S Rathi, Nathan M Kendsersky, Michael Randall, Laura Donovan, Sorana Morrissy, Robyn T Sussman, Doncho V Zhelev, Yang Feng, Yanping Wang, Jennifer Hwang, Gonzalo Lopez, Jo Lynne Harenza, Jun S Wei, Bruce Pawel, Tricia Bhatti, Mariarita Santi, Arupa Ganguly, Javed Khan, Marco A Marra, Michael D Taylor, Dimiter S Dimitrov, Crystal L Mackall, John M Maris
ABSTRACT
We developed an RNA-sequencing-based pipeline to discover differentially expressed cell-surface molecules in neuroblastoma that meet criteria for optimal immunotherapeutic target safety and efficacy. Here, we show that GPC2 is a strong candidate immunotherapeutic target in this childhood cancer. We demonstrate high GPC2 expression in neuroblastoma due to MYCN transcriptional activation and/or somatic gain of the GPC2 locus. We confirm GPC2 to be highly expressed on most neuroblastomas, but not detectable at appreciable levels in normal childhood tissues. In addition, we demonstrate that GPC2 is required for neuroblastoma proliferation. Finally, we develop a GPC2-directed antibody-drug conjugate that is potently cytotoxic to GPC2-expressing neuroblastoma cells. Collectively, these findings validate GPC2 as a non-mutated neuroblastoma oncoprotein and candidate immunotherapeutic target.
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.